missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2,2-Trifluoroethoxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15439825
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439825 250 mg 250mg
15459217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439825 Supplier Thermo Scientific Alfa Aesar Supplier No. H52471.MD
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 957060-90-1 | Molecular Formula: C8H8BF3O3 | Molecular Weight: 219.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957060-90-1
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD09027258
InChI Key IFWDRBWGMUCFMH-UHFFFAOYSA-N
Synonym 2-2,2,2-trifluoroethoxy phenylboronic acid, 2-2,2,2-trifluoroethoxy benzeneboronic acid, 2-2,2,2-trifluoroethoxy phenyl boronic acid, acmc-209s1a, 2-2,2,2-trifluoroethoxy phenyl boronicacid
PubChem CID 43145273
IUPAC Name [2-(2,2,2-trifluoroethoxy)phenyl]boronic acid
SMILES B(C1=CC=CC=C1OCC(F)(F)F)(O)O

Specifications

Melting Point 125°C to 128°C
Quantity 250 mg
Formula Weight 219.95
Percent Purity 98%
Chemical Name or Material 2-(2,2,2-Trifluoroethoxy)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.