missing translation for 'onlineSavingsMsg'
Learn More

2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride, TRC

Product Code. 30382752
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382752 10 mg
30369457 50 mg
30378874 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382752 Supplier Toronto Research Chemicals Supplier No. TRCD41868610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1825392-15-1
Molecular Formula C25H33N3O4S.2HCl
Molecular Weight (g/mol) 544.53
Synonym 2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride, Ethanol, 2-[2-[2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethoxy]ethoxy]-, hydrochloride (1:2) (ACI)
IUPAC Name 2-[2-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethoxy]ethoxy]ethanol;dihydrochloride
SMILES Cl.Cl.OCCOCCOCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H33N3O4S.2ClH/c29-14-16-31-18-20-32-19-17-30-15-13-27-9-11-28(12-10-27)25-21-5-1-3-7-23(21)33-24-8-4-2-6-22(24)26-25;;/h1-8,29H,9-20H2;2*1H
Formula Weight 543.1725
Chemical Name or Material 2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.