missing translation for 'onlineSavingsMsg'
Learn More

2-[(1RS)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Methyl Etodolac), Mikromol™

Product Code. 30049807
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049807 25 mg 25mg
18093010 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049807 Supplier Mikromol Supplier No. MM0911.030025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[(1RS)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Methyl Etodolac)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methyl Etodolac
CAS 109518-47-0
Analyte or Component Names 1-Methyl Etodolac
Shipping Condition Room Temperature
Molecular Formula C16 H19 N O3
Certifications/Compliance ISO 17025
Synonym Etodolac Imp. C (EP), Etodolac USP Related Compound A, Etodolac USP RC A, 2-[(1RS)-8-Ethyl-1-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, 1-Methyl Etodolac, Etodolac Related Compound A, Etodolac Impurity C
InChI Formula InChI=1S/C16H19NO3/c1-3-10-5-4-6-11-12-7-8-20-16(2,9-13(18)19)15(12)17-14(10)11/h4-6,17H,3,7-9H2,1-2H3,(H,18,19)
SMILES CCc1cccc2c3CCOC(C)(CC(=O)O)c3[nH]c12
IUPAC Name 2-(8-ethyl-1-methyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
Molecular Weight (g/mol) 273.33
Quantity 25 mg
Formula Weight 273.1365 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.