missing translation for 'onlineSavingsMsg'
Learn More

2-[(1RS)-3-(methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Mikromol

Product Code. 30463181
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30463181 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463181 Supplier Mikromol Supplier No. MM1122.100025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[(1RS)-3-(methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Mikromol
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material 2-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
CAS 1033803-59-6
Analyte or Component Names 2-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
Shipping Condition Room Temperature
Molecular Formula C18H19NOS
Certifications/Compliance ISO 9001
Synonym 1-Naphthalenol, 2-[3-(methylamino)-1-(2-thienyl)propyl]-, 2-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol, 2-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Duloxetine Hydrochloride Imp. E (EP)
InChI Formula InChI=1S/C18H19NOS/c1-19-11-10-15(17-7-4-12-21-17)16-9-8-13-5-2-3-6-14(13)18(16)20/h2-9,12,15,19-20H,10-11H2,1H3
Solution Type Neat
SMILES CNCCC(c1cccs1)c2ccc3ccccc3c2O
IUPAC Name 2-[3-(methylamino)-1-(2-thienyl)propyl]naphthalen-1-ol
Molecular Weight (g/mol) 297.41
Quantity 25 mg
Formula Weight 297.1187 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.