missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac), Mikromol™
Description
2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac)
Specifications
Specifications
| Chemical Name or Material | 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac) |
| CAS | 41339-67-7 |
| Analyte or Component Names | 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H17 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Etodolac Imp. A (EP), 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, 8-Desethyl Etodolac, Etodolac Impurity A |
| InChI Formula | InChI=1S/C15H17NO3/c1-2-15(9-13(17)18)14-11(7-8-19-15)10-5-3-4-6-12(10)16-14/h3-6,16H,2,7-9H2,1H3,(H,17,18) |
| SMILES | CCC1(CC(=O)O)OCCc2c1[nH]c3ccccc23 |
| IUPAC Name | 2-(1-ethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?