missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-indol-3-yl)ethan-1-ol, Thermo Scientific™

Product Code. 10720524
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10720524 10 g Amber Glass Bottle 10g
10056192 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10720524 Supplier Thermo Scientific Maybridge Supplier No. BTB14321EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
IUPAC Name 2-(1H-indol-3-yl)ethanol
SMILES OCCC1=CNC2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Merck Index 15, 9978
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.