missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Benzimidazol-1-yl)acetic acid, 90%, Thermo Scientific™

Product Code. 10285854
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285854 250 mg Amber Glass Bottle 250mg
10061834 1 g Amber Glass Bottle 1g
10012464 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10285854 Supplier Thermo Scientific Maybridge Supplier No. MO01258CB
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 40332-16-9
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
InChI Key AFHSZBKDUTWXDX-UHFFFAOYSA-N
Synonym benzoimidazol-1-yl-acetic acid, 1h-benzimidazole-1-acetic acid, 1h-benzimidazol-1-ylacetic acid, 2-1h-benzo d imidazol-1-yl acetic acid, 2-1h-1,3-benzodiazol-1-yl acetic acid, benzoimidazol-1-ylacetic acid, 1h-benzimidazole-1-aceticacid, 1,3-benzodiazol-1-ylacetic acid, 2-benzimidazol-1-yl acetic acid, 1h-benzimidazol-1-yl acetic acid
PubChem CID 805645
IUPAC Name 2-(benzimidazol-1-yl)acetic acid
SMILES C1=CC=C2C(=C1)N=CN2CC(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.17
Percent Purity 90%
Chemical Name or Material 2-(1H-Benzimidazol-1-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.