missing translation for 'onlineSavingsMsg'
Learn More

2-(1h-1,2,4-Triazol-1-yl)acetic Acid, TRC

Product Code. 30137162
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137162 500 mg 500mg
30133717 1 g 1g
30003865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137162 Supplier Toronto Research Chemicals Supplier No. TRCT898595500MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28711-29-7
Molecular Formula C4 H5 N3 O2
Molecular Weight (g/mol) 127.1
Synonym 1H-1,2,4-Triazole-1-acetic acid, 1,2,4-Triazol-1-ylacetic acid, 1,2,4-Triazole-1-acetic acid, 1H-1,2,4-Triazol-1-ylacetic acid, 2-(1,2,4-Triazol-1-yl)acetic acid, 2-(1H-1,2,4-Triazol-1-yl)acetic acid, (1,2,4-Triazol-1-yl)acetic acid
IUPAC Name 2-(1,2,4-triazol-1-yl)acetic acid
SMILES OC(=O)Cn1cncn1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H5N3O2/c8-4(9)1-7-3-5-2-6-7/h2-3H,1H2,(H,8,9)
Formula Weight 127.0382
Percent Purity >95%
Chemical Name or Material (1,2,4-Triazol-1-yl) acetic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.