missing translation for 'onlineSavingsMsg'
Learn More

2-[(11bS)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™

Product Code. 15425465
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425465 250 mg 250mg
15415465 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425465 Supplier Thermo Scientific Alfa Aesar Supplier No. H60324.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1223007-01-9
Molecular Formula C24H23BF4NP
Molecular Weight (g/mol) 443.232
MDL Number MFCD17018773
InChI Key AMEHGPFCNMMXTB-UHFFFAOYSA-O
Synonym 2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborate
PubChem CID 72376450
SMILES [B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54

Especificaciones

Quantity 250 mg
Formula Weight 443.22
Percent Purity ≥97%
Chemical Name or Material 2-[(11bS)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.