missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)phenol, 98%

Product Code. 11368025
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368025 5 g 5g
11358025 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368025 Supplier Thermo Scientific Alfa Aesar Supplier No. L08178.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4787-77-3 | C10H13NO | 163.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4787-77-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00051659
InChI Key ZGSBDRFDXWRZAE-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl phenol, 2-pyrrolidinophenol, 2-1-pyrrolidino phenol, 2-1-pyrrolidinyl phenol, 2-tetrahydro-1h-pyrrol-1-ylphenol, 2-pyrrolidinylphenol, 2-pyrrolizinophenol, 2-pyrrolidin-1-yl-phenol, phenol, 2-1-pyrrolidinyl, 1-2-hydroxyphenyl pyrrolidine
PubChem CID 2734845
IUPAC Name 2-pyrrolidin-1-ylphenol
SMILES OC1=CC=CC=C1N1CCCC1

Specifications

Melting Point 109°C to 112°C
Quantity 5 g
Beilstein 135437
Solubility Information Insoluble in water.
Formula Weight 163.22
Percent Purity 98%
Chemical Name or Material 2-(1-Pyrrolidinyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.