missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)nicotinic acid, 95%, Thermo Scientific™

Product Code. p-3075324
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10759173 250 mg 250mg
10255334 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10759173 Supplier Thermo Scientific Maybridge Supplier No. MO01300CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-36-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD05865128
InChI Key DTUSJCJZQJAPGD-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl nicotinic acid, 2-pyrrolidin-1-yl pyridine-3-carboxylic acid, 2-pyrrolidin-1-ylnicotinic acid, 2-pyrrolidin-1-yl nicotinic acid, 2-pyrrolidinylpyridine-3-carboxylic acid, 2-1-pyrrolidinyl nicotinicacid, 3-pyridinecarboxylic acid, 2-1-pyrrolidinyl
PubChem CID 2794739
SMILES OC(=O)C1=C(N=CC=C1)N1CCCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2-(pyrrolidin-1-yl)pyridine-3-carboxylic acid
Formula Weight 192.22
Percent Purity 95%
Chemical Name or Material 2-(1-Pyrrolidinyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.