missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)benzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 11409363
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72752-52-4
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD00049221
InChI Key MEBVSLLKZSAIGK-UHFFFAOYSA-N
Synonym 2-piperidinobenzonitrile, 2-piperidin-1-yl benzonitrile, 2-1-piperidinyl benzonitrile, gnf-pf-3161, benzonitrile, 2-1-piperidinyl, 2-piperidylbenzenecarbonitrile, 2-piperidino benzonitrile, maybridge1_000930, acmc-1bfz1, 2-1-piperidyl benzonitrile
PubChem CID 2774355
IUPAC Name 2-piperidin-1-ylbenzonitrile
SMILES C1CCN(CC1)C2=CC=CC=C2C#N

Specifications

Melting Point 41°C to 45°C
Boiling Point 122°C to 125°C (1 mmHg)
Quantity 1 g
UN Number UN3077
Beilstein 1243909
Formula Weight 186.26
Percent Purity 97%
Chemical Name or Material 2- (1-Piperidinyl)benzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'