missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)benzonitrile, 97%

Product Code. 11419363
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419363 5 g 5g
11409363 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419363 Supplier Thermo Scientific Alfa Aesar Supplier No. A11784.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72752-52-4 | C12H14N2 | 186.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72752-52-4
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD00049221
InChI Key MEBVSLLKZSAIGK-UHFFFAOYSA-N
Synonym 2-piperidinobenzonitrile, 2-piperidin-1-yl benzonitrile, 2-1-piperidinyl benzonitrile, gnf-pf-3161, benzonitrile, 2-1-piperidinyl, 2-piperidylbenzenecarbonitrile, 2-piperidino benzonitrile, maybridge1_000930, acmc-1bfz1, 2-1-piperidyl benzonitrile
PubChem CID 2774355
IUPAC Name 2-piperidin-1-ylbenzonitrile
SMILES C1CCN(CC1)C2=CC=CC=C2C#N

Specifications

Melting Point 41°C to 45°C
Boiling Point 122°C to 125°C (1 mmHg)
Quantity 5 g
UN Number UN3077
Beilstein 1243909
Formula Weight 186.26
Percent Purity 97%
Chemical Name or Material 2- (1-Piperidinyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.