missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)pyrimidine dihydrochloride, 98+%, Thermo Scientific™

Product Code. 10667572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667572 5 g Glass bottle 5g
10249213 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667572 Supplier Thermo Scientific Acros Supplier No. 243290050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94021-22-4
MDL Number MFCD00012779
InChI Key ZNZGJSLHXOMREP-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 1-2-pyrimidinyl piperazine dihydrochloride, 2-piperazinylpyrimidine hcl, 2-piperazinylpyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride, pubchem7349
PubChem CID 11680272
IUPAC Name 2-piperazin-1-ylpyrimidine;dihydrochloride
SMILES C1CN(CCN1)C2=NC=CC=N2.Cl.Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 286.0°C to 288.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C8H14Cl2N4
Quantity 5 g
Molecular Weight (g/mol) 237.128
Formula Weight 237.13
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-(1-Piperazinyl)pyrimidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.