missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)nicotinamide, 95%

Product Code. p-7033797
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499720 250 mg 250mg
11409730 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499720 Supplier Thermo Scientific Alfa Aesar Supplier No. H50967.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87394-64-7 | C10H14N4O | 206.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87394-64-7
Molecular Formula C10H14N4O
Molecular Weight (g/mol) 206.25
MDL Number MFCD08061093
InChI Key RYNARQDUUQTTFJ-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl nicotinamide, 2-piperazin-1-ylnicotinamide, 2-1-piperazinyl nicotinamide, 2-piperazin-1-yl pyridine-3-carboxamide, 2-piperazin-1-yl-nicotinamide, chembl47688, 2-piperazin-1-ylnicotinamide dihydrochloride, acmc-1bjdr, 2-1-piperazino nicotinamide, 1-3-carbamoyl-2-pyridyl piperazine
PubChem CID 16785637
IUPAC Name 2-piperazin-1-ylpyridine-3-carboxamide
SMILES NC(=O)C1=C(N=CC=C1)N1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 206.25
Percent Purity 95%
Chemical Name or Material 2-(1-Piperazinyl)nicotinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.