missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthylmethyl)-2-imidazoline nitrate, 99%

Product Code. 11383266
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383266 100 g 100g
11373266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383266 Supplier Thermo Scientific Alfa Aesar Supplier No. L12617.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5144-52-5 | C14H15N3O3 | 273.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5144-52-5
Molecular Formula C14H15N3O3
Molecular Weight (g/mol) 273.292
MDL Number MFCD00014316
InChI Key ZAHXYMFVNNUHCP-UHFFFAOYSA-N
Synonym naphazoline nitrate, privine, naphcon, 2-1-naphthylmethyl-2-imidazoline nitrate, unii-sc99gr1t5s, naphthisen, naphthizen, vistalbalon, imidazyl, naftizin
PubChem CID 82332
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;nitric acid
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.[N+](=O)(O)[O-]

Specifications

Melting Point 168°C to 170°C
Quantity 100 g
Beilstein 3779329
Sensitivity Hygroscopic
Formula Weight 273.29
Percent Purity 99%
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.