missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanol, TRC

Product Code. 30474992
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474992 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474992 Supplier Toronto Research Chemicals Supplier No. TRCN37832510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(1-Naphthyl)ethanol
CAS 773-99-9
Molecular Formula C12 H12 O
Quantity 10 g
InChI Formula InChI=1S/C12H12O/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7,13H,8-9H2
SMILES OCCc1cccc2ccccc12
IUPAC Name 2-naphthalen-1-ylethanol
Molecular Weight (g/mol) 172.22
Formula Weight 172.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.