missing translation for 'onlineSavingsMsg'
Learn More

2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzaldehyde, 97%, Thermo Scientific™

Product Code. 10092204
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092204 1 g Amber Glass Bottle 1g
10471884 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10092204 Supplier Thermo Scientific Maybridge Supplier No. CC62104DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-61-7
Molecular Formula C12H9F3N2O
Molecular Weight (g/mol) 254.21
MDL Number MFCD09817510
InChI Key STBRCNYURIXHAU-UHFFFAOYSA-N
Synonym 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzaldehyde, 2-2-methyl-5-trifluoromethyl pyrazol-3-yl benzaldehyde, 2-1-methyl-3-trifluoromethyl pyrazol-5-yl benzaldehyde, 2-2-methyl-5-trifluoromethyl-3-pyrazolyl benzaldehyde, 5-2-formylphenyl-1-methyl-3-trifluoromethyl-1h-pyrazole, benzaldehyde,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl
PubChem CID 24229657
IUPAC Name 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzaldehyde
SMILES CN1N=C(C=C1C1=CC=CC=C1C=O)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 254.21
Percent Purity 97%
Chemical Name or Material 2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzaldehyde
compliance-icons
Product Identifier
  • 2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.