missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methyl-1H-imidazol-5-yl)acetic Acid Hydrochloride, TRC

Product Code. 30473040
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473040 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473040 Supplier Toronto Research Chemicals Supplier No. TRCM3229082.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(1-Methyl-1H-imidazol-5-yl)acetic Acid Hydrochloride
CAS 1071661-55-6
Molecular Formula C6 H8 N2 O2 . H Cl
Quantity 2.5 mg
InChI Formula InChI=1S/C6H8N2O2.ClH/c1-8-4-7-3-5(8)2-6(9)10;/h3-4H,2H2,1H3,(H,9,10);1H
SMILES Cl.Cn1cncc1CC(=O)O
IUPAC Name 2-(3-methylimidazol-4-yl)acetic acid;hydrochloride
Molecular Weight (g/mol) 176.601
Formula Weight 176.035
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.