missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Cyclohexenyl)cyclohexanone, 85+%, cont. ca 10% 2-cyclohexylidenecyclohexanone

Product Code. 11392267
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11392267

Brand: Thermo Scientific Alfa Aesar L09798.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1502-22-3 | C12H18O | 178.28 g/mol

It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1502-22-3
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00013796
InChI Key GVNVAWHJIKLAGL-UHFFFAOYNA-N
Synonym 2-1-cyclohexenyl cyclohexanone, chch, 2-cyclohexenylcyclohexanone, 2-1-cyclohexen-1-yl cyclohexan-1-one, 1,1'-bi cyclohexan-1-en-2-one, cyclohexanone, 2-1-cyclohexen-1-yl, 2-cyclohex-1-enyl cyclohexanone, 2-cyclohex-1-en-1-yl cyclohexan-1-one, 2-1-cyclohexen-1-yl cyclohexanone, cyclohexanone, 2-cyclohexenyl
PubChem CID 101175
SMILES O=C1CCCCC1C1=CCCCC1

Specifications

Density 0.998
Boiling Point 265°C
Flash Point >110°C (230°F)
Refractive Index 1.506
Quantity 25 g
Beilstein 1308501
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name 2-(cyclohexen-1-yl)cyclohexan-1-one
Formula Weight 178.28
Percent Purity ≥85%
Assay cont. ≈10% 2-cyclohexylidenecyclohexanone
Chemical Name or Material 2-(1-Cyclohexenyl)cyclohexanone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.