missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Adamantyl)acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Spécification
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 25 g |
IUPAC Name | 2-(1-adamantyl)acetic acid |
Formula Weight | 194.27 |
Percent Purity | 97% |
Chemical Name or Material | 2-(1-Adamantyl)acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'