missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonate Potassium, TRC

Product Code. 30376287
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376287 1 g
30380142 500 mg
30382835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376287 Supplier Toronto Research Chemicals Supplier No. TRCD4879331G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91893-72-0
Molecular Formula C10H8KNO5S
Molecular Weight (g/mol) 293.34
SMILES [K+].[O-]S(=O)(=O)CCN1C(=O)c2ccccc2C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H9NO5S.K/c12-9-7-3-1-2-4-8(7)10(13)11(9)5-6-17(14,15)16;/h1-4H,5-6H2,(H,14,15,16);/q;+1/p-1
Formula Weight 292.976
Chemical Name or Material 2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)ethanesulfonate Potassium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.