missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazol-4-amine, 97%, Thermo Scientific™

Product Code. 10532432
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10532432 10 g 10g
10170572 1 g 1g
10784277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10532432 Supplier Thermo Scientific Maybridge Supplier No. BTB13048EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 767-63-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.126
MDL Number MFCD00168448
InChI Key IPCMVRZVNJHUHR-UHFFFAOYSA-N
Synonym 4-aminobenzofurazan, benzo 1,2,5 oxadiazol-4-ylamine, 2,1,3-benzooxadiazol-4-ylamine, benzo c 1,2,5-oxadiazole-4-ylamine, benzofurazan-4-amine, benzofurazane-4-amine, acmc-209p5u, 2,1,3-benzoxadiazol-7-amine, 2,1,3-benzoxadiazol-4-ylamine, benzo c 1,2,5 oxadiazol-4-amine
PubChem CID 584570
IUPAC Name 2,1,3-benzoxadiazol-4-amine
SMILES C1=CC2=NON=C2C(=C1)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 135.13
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.