missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Product Code. 11390338
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390338 2 g 2g
11380338 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390338 Supplier Thermo Scientific Alfa Aesar Supplier No. 043730.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The product is used as an industrial solvent, and pharmaceutical intermediates for organic synthesis and used nitrocellulose and other resin, wax, fatty, dyes and other solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-13-2
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00005809
InChI Key PDQRQJVPEFGVRK-UHFFFAOYSA-N
Synonym piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
PubChem CID 67502
IUPAC Name 2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1

Specifications

Melting Point 42°C to 44°C
Boiling Point 206°C
Flash Point 95°C (203°F)
Quantity 2 g
Formula Weight 136.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,1,3-Benzothiadiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.