missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10214934
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10214934 5 g 5g
10344472 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214934 Supplier Thermo Scientific Maybridge Supplier No. CC09103DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 73713-79-8
Molecular Formula C6H3ClN2O2S2
Molecular Weight (g/mol) 234.67
MDL Number MFCD00068049
InChI Key CXAICGCTHOWKPP-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
PubChem CID 2735459
IUPAC Name 2,1,3-benzothiadiazole-4-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC2=NSN=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 234.69
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.