Learn More
2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Maybridge™
Brand: Maybridge CC09104DA
Code : 31
Additional Details : CAS Number : 5170-68-3 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
5170-68-3 | |
164.182 | |
ANVJARPTPIVPRC-UHFFFAOYSA-N | |
2776283 | |
C1=CC2=NSN=C2C(=C1)C=O |
C7H4N2OS | |
MFCD02681902 | |
benzo c 1,2,5 thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carboxaldehyde, tos-bb-1233, 2,1,3-benzothiadiazol-4-aldehyde, 2,1,3-benzothiadiazole-4-aldehyde, 2,1,3-benzothiadiazol-4-carboxaldehyde, benzo c 1,2,5-thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carbaldehyde | |
2,1,3-benzothiadiazole-4-carbaldehyde |
Specifications
2, 1, 3-Benzothiadiazole-4-carbaldehyde | |
1g | |
94.5 | |
95% | |
MFCD02681902 | |
ANVJARPTPIVPRC-UHFFFAOYSA-N | |
2,1,3-benzothiadiazole-4-carbaldehyde | |
2776283 | |
95% |
Amber Glass Bottle | |
5170-68-3 | |
100.0 | |
C7H4N2OS | |
benzo c 1,2,5 thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carboxaldehyde, tos-bb-1233, 2,1,3-benzothiadiazol-4-aldehyde, 2,1,3-benzothiadiazole-4-aldehyde, 2,1,3-benzothiadiazol-4-carboxaldehyde, benzo c 1,2,5-thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carbaldehyde | |
C1=CC2=NSN=C2C(=C1)C=O | |
164.182 | |
164.19 |