missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazol-4-yl isocyanate, 97%, Thermo Scientific™

Product Code. 10113454
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10113454 5 g 5g
10763233 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113454 Supplier Thermo Scientific Maybridge Supplier No. CC09106DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 342411-14-7
Molecular Formula C7H3N3OS
Molecular Weight (g/mol) 177.181
MDL Number MFCD02681903
InChI Key HPZZFYLZFYDNMA-UHFFFAOYSA-N
PubChem CID 2776284
IUPAC Name 4-isocyanato-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C(=C1)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 2,1,3-benzothiadiazol-4-yl isocyanate, 2,1,3-benzothiadiazol-4-yl isoxyanate, 4-isocyanatobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazol-4-ylisocyanate, 2,1,3-benzothiadiazole,4-isocyanato, benzo c 1,2,5-thiadiazol-4-isocyanate, 2,1,3-benzothiadiazole, 4-isocyanato, 2,1,3-benzothiadiazole, 4-isocyanato-9ci
Formula Weight 177.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazol-4-yl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.