missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Benzodioxol-5-yl)ethanol, ≥97%, Thermo Scientific™

Product Code. 10072414
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10072414 1 g 1g
10432504 250 mg 250mg
10629315 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10072414 Supplier Thermo Scientific Maybridge Supplier No. CC50809DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6006-82-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00666033
InChI Key JADSGOFBFPTCHG-UHFFFAOYSA-N
Synonym 2-benzo d 1,3 dioxol-5-yl ethanol, 2-2h-1,3-benzodioxol-5-yl ethan-1-ol, 3,4-methylenedioxy phenethyl alcohol, 2-1,3-benzodioxol-5-yl ethanol, 1,3-benzodioxole-5-ethanol, homopiperonyl alcohol, 3,4-methylenedioxyphenethyl alcohol, 2-1,3-benzodioxol-5-yl-ethanol, 2-2h-1,3-benzodioxol-5-yl ethanol, benzodioxolylethanol
PubChem CID 2759846
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanol
SMILES C1OC2=C(O1)C=C(C=C2)CCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2-(1, 3-Benzodioxol-5-yl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.