missing translation for 'onlineSavingsMsg'
Learn More

2-(1,2-Benzisoxazol-3-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10220595
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10220595 5 g 5g
10425712 250 mg 250mg
10425902 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10220595 Supplier Thermo Scientific Maybridge Supplier No. KM10833DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4865-84-3
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD02180388
InChI Key BVSIAYQIMUUCRW-UHFFFAOYSA-M
Synonym 2-1,2-benzisoxazol-3-yl acetic acid, 1,2-benzisoxazole-3-acetic acid, 1,2-benzisoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl acetic acid, 2-benzo d isoxazol-3-yl acetic acid, 1,2-benzoxazol-3-ylacetic acid, 2-benzo d isoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl ethanoic acid, nvu, zlchem 388
PubChem CID 301729
SMILES [O-]C(=O)CC1=NOC2=CC=CC=C12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 2-(1,2-benzoxazol-3-yl)acetic acid
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 2-(1, 2-Benzisoxazol-3-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.