missing translation for 'onlineSavingsMsg'
Learn More

2-(1,1-Difluoroethyl)pyrazine, 97%, Thermo Scientific™

Product Code. 15249756 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15249756 5 g 5g
15239756 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15249756 Supplier Thermo Scientific Acros Supplier No. 451360050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 111781-49-8
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.12
InChI Key OXKFWGSACYGCSY-UHFFFAOYSA-N
Synonym 2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl
PubChem CID 19763564
IUPAC Name 2-(1,1-difluoroethyl)pyrazine
SMILES CC(C1=NC=CN=C1)(F)F

Specifications

Boiling Point 150.0°C to 152.0°C
Flash Point 53°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2-(1, 1-Difluoroethyl)pyrazine
compliance-icons compliance-icons
Product Identifier
  • 2-(1,1-Difluoroethyl)pyrazine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.