missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences 1x Buffer XT-R - Regeneration Buffer

Product Code. 16654714 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 x 250 mL
Unit Size:
1 bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16654714 1 x 250 mL 1 bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16654714 Supplier IBA Lifesciences Supplier No. 21045250

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Buffer XT-R is a Strep-Tactin™XT regeneration buffer with magnesium chloride

  • Buffer XT-R is a ready-to-use Strep-Tactin™XT regeneration buffer containing magnesium chloride for competitive displacement of biotin from the modified biotin-binding pocket of Strep-Tactin™XT after the elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • Please note that Buffer XT-R is not applicable for Strep-Tactin™.

Specifications

Description Ready-to-use
Concentration 10X
For Use With (Application) Protein Purification
Physical Form Solution
Quantity 1 x 250 mL
Solution Type Buffer
Formulation Solution
Grade Molecular Biology

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.