Learn More
Merck 1SET Schaeffer and Fulton Spore Stain Kit
Brand: Merck 04551-1KT-F
1SET Schaeffer and Fulton Spore Stain Kit
Description
Safety and Handling
- Schaeffer and Fulton Spore Stain Kit
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.