missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A

Product Code. 18769558
Click to view available options
Quantity:
2 Plex
5 Plex
7 Plex
Unit Size:
2 sets
5 sets
7 sets
This item is not returnable. View return policy

Product Code. 18769558

Brand: R&D Systems™ FCSTM1302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Earn double rewards for reviews

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 Plex
Species Human
Binding Target Multiplex Cytokine Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.