missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Verbenone, 94%

Product Code. 10441441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441441 50 g Glass bottle 50g
10522611 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441441 Supplier Thermo Scientific Acros Supplier No. 203630500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1196-01-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 227°C to 228°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.497
Quantity 50 g
Beilstein 07, 161
Specific Gravity 0.97
Specific Rotation Condition − 220.00 (20.00°C c=3−4,CHCL3)
Specific Rotation -220
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with most common organic solvents
Formula Weight 150.22
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material (1S)-(-)-Verbenone, 94%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.