missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1S)-(+)-Menthyl glyoxylate monohydrate, 98%

Product Code. 11359745
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359745 25 g 25g
11349745 5 g 5g
11369745 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359745 Supplier Alfa Aesar™ Supplier No. L19785.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C12H22O4
MDL Number MFCD04037548
Synonym Glyoxylic acid (1S)-menthyl ester hydrate

Specifications

Melting Point 76°C to 80°C
Density 1.182
Boiling Point 95°C to 96°C (0.3 mmHg)
Quantity 25 g
UN Number UN3077
Beilstein 4905777
Sensitivity Air sensitive
Optical Rotation +74° (c=1 in Methanol)
Formula Weight 230.31 (212.29 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1S)-(+)-Menthyl glyoxylate monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.