missing translation for 'onlineSavingsMsg'
Learn More

(1S)-Camphorsultam, TRC

Product Code. 30334975
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334975 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30334975 Proveedor Toronto Research Chemicals N.º de proveedor TRCC1751252G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S)-Camphorsultam
CAS 94594-90-8
Molecular Formula C10H17NO2S
Quantity 2 g
Synonym (1S)-(-)-10,2-Camphorsultam, (2R)-Bornane-10,2-sultam, Hexahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole
InChI Formula InChI=1S/C10H17NO2S/c1-9(2)7-3-4-10(9)6-14(12,13)11-8(10)5-7/h7-8,11H,3-6H2,1-2H3/t7-,8-,10-/m1/s1
SMILES CC1(C)[C@@H]2CC[C@]13CS(=O)(=O)N[C@@H]3C2
IUPAC Name (5R)-10,10-dimethyl-3λ6-thia-4-azatricyclo[5.2.1.01,5]decane 3,3-dioxide
Molecular Weight (g/mol) 215.13
Formula Weight 215.098
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.