missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Camphorsulfonylimine, 98+%, Thermo Scientific Chemicals

Catalog Number 11415756
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
60886-80-8
C10H15NO2S
213.30
MFCD00013315,MFCD00064576
ZAHOEBNYVSWBBW-UHFFFAOYNA-N
1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione, 1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione
45925640
10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
CC1(C)C2CCC11CS(=O)(=O)N=C1C2
This item is not returnable. View return policy
60886-80-8
C10H15NO2S
213.30
MFCD00013315,MFCD00064576
ZAHOEBNYVSWBBW-UHFFFAOYNA-N
1s,7s-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.0<1,5> dec-4-ene-3,3-dione, 1s,7s-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? dec-4-ene-3,3-dione
45925640
10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
CC1(C)C2CCC11CS(=O)(=O)N=C1C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 226°C to 228°C
Quantity 1 g
Beilstein 85296
Formula Weight 213.3
Percent Purity ≥98%
Chemical Name or Material (1S)-(-)-Camphorsulfonylimine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'