missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1S)-(+)-Camphor-10-sulfonyl chloride, 97%

Product Code. 11400873
Change view
Click to view available options
Quantity:
5g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400873 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400873 Supplier Alfa Aesar™ Supplier No. A13293.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21286-54-4
Molecular Formula C10H15ClO3S
MDL Number MFCD00064156
Quantity 5g
UN Number UN3261
Beilstein 3205974
Synonym d +-10-camphorsulfonyl chloride, 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonyl chloride, 1s-+-camphor-10-sulfonyl chloride, d-2-oxobornane-10-sulphonyl chloride, 1s-+-10-camphorsulfonyl chloride, d-+-10-camphorsulfonyl chloride, camphor-10-sulfonyl chloride, +-camphor-10-sulfonyl chloride, +-camphor-10-sulfonic acid chloride
InChI Key BGABKEVTHIJBIW-GMSGAONNSA-N
Optical Rotation +32° (c=1 in Chloroform)
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)Cl)C
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonyl chloride
Molecular Weight (g/mol) 250.737
PubChem CID 88856
Formula Weight 250.74
Percent Purity 97%
Chemical Name or Material (1S)-(+)-Camphor-10-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.