missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Camphanic chloride, 95%

Product Code. 11329005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329005 1 g 1g
11339005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329005 Supplier Thermo Scientific Alfa Aesar Supplier No. L14148.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39637-74-6 | C10H13ClO3 | 216.661 g/mol

(1S)-(-)-Camphanic chloride is used as a resolving agent for alcohols as the diastereomeric esters by crystallization or chromatography and as a chiral derivatization reagent for determination of enantiomeric excess of alcohols and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(-)-Camphanic chloride is used as a resolving agent for alcohols as the diastereomeric esters by crystallization or chromatography and as a chiral derivatization reagent for determination of enantiomeric excess of alcohols and amines.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to moisture. Protect from humidity and water. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39637-74-6
Molecular Formula C10H13ClO3
Molecular Weight (g/mol) 216.661
MDL Number MFCD00135626
InChI Key PAXWODJTHKJQDZ-QVDQXJPCSA-N
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
PubChem CID 12364938
IUPAC Name (4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
SMILES CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C

Specifications

Melting Point 64°C to 68°C
Quantity 1 g
UN Number UN3261
Beilstein 3590860
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol. Reacts with water.
Optical Rotation −18° (c=2 in Carbon Tetrachloride)
Formula Weight 216.67
Percent Purity 95%
Chemical Name or Material (1S)-(-)-Camphanic chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.