missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Melting Point | 152°C |
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Packaging | Glass Bottle |
| Solubility Information | Solubility in water: soluble |
| Quantity | 1 g |
| Formula Weight | 227.26 |
| Show More |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?