missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™

Product Code. 10095073 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095073 250 mg Glass Bottle 250mg
10394252 1 g Glass Bottle 1g
10098751 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095073 Supplier Thermo Scientific Acros Supplier No. 361822500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 151907-80-1
Molecular Formula C11H17NO4
Molecular Weight (g/mol) 227.26
MDL Number MFCD00211288
InChI Key WOUNTSATDZJBLP-UHFFFAOYNA-N
Synonym 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid
PubChem CID 7019281
IUPAC Name (1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O

Specifications

Melting Point 152°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
Quantity 250 mg
Formula Weight 227.26
Percent Purity 95%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (1S, 4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 98% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.