missing translation for 'onlineSavingsMsg'
Learn More

(-)-(1S,4R)-Camphanoyl Chloride, TRC

Product Code. 30347048
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347048 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347048 Supplier Toronto Research Chemicals Supplier No. TRCC1650005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (-)-(1S,4R)-Camphanoyl Chloride
CAS 39637-74-6
Molecular Formula C10H13ClO3
Quantity 5 g
InChI Formula InChI=1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m0/s1
SMILES CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(=O)Cl
IUPAC Name (1R,4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
Molecular Weight (g/mol) 216.66
Formula Weight 216.055
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.