missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-(1S,4R)-Camphanoyl Chloride, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | (-)-(1S,4R)-Camphanoyl Chloride |
| CAS | 39637-74-6 |
| Molecular Formula | C10H13ClO3 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m0/s1 |
| SMILES | CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(=O)Cl |
| IUPAC Name | (1R,4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride |
| Molecular Weight (g/mol) | 216.66 |
| Formula Weight | 216.055 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction