missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(1S,4R)-7,7-Dimethyl-2-oxo-bicyclo[2.2.1]hept-1-yl]methanesulfonic Acid ((+)-10-Camphorsulfonic Acid), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
[(1S,4R)-7,7-Dimethyl-2-oxo-bicyclo[2.2.1]hept-1-yl]methanesulfonic Acid ((+)-10-Camphorsulfonic Acid)
Specifications
Specifications
| Chemical Name or Material | (1S)-(+)-10-Camphorsulfonic Acid |
| CAS | 3144-16-9 |
| Analyte or Component Names | (1S)-(+)-10-Camphorsulfonic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H16 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1S,4R)-, 10-Bornanesulfonic acid, 2-oxo-, (1S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, (1S)-, Camphorsulfonic acid (6CI), (+)-(S)-Camphor-10-sulfonic acid, (+)-2-Oxo-10-bornanesulfonic acid, (+)-Camphor-10-sulfonic acid, (+)-Camphor-ω-sulfonic acid, (+)-Camphorsulfonic acid, (+)-D-Camphor-10-sulfonic acid, (+)-β-Camphorsulfonic acid, (1S)-(+)-10-Camphorsulfonic acid, (1S)-10-Camphorsulfonic acid, (S)-(+)-CSA, (S)-(+)-Camphor-10-sulfonic acid, (S)-Camphor-10-sulfonic acid, 2-Oxo-10-bornanesulfonic acid, Camphor-10-sulfonic acid, (+)-, Camphor-ω-sulfonic acid, (+)-, D-(+)-Camphor-10-sulfonic acid, D-(+)-Camphorsulfonic acid, D-Camphor-10-sulfonic acid, D-Camphor-β-sulfonic acid, D-Camphorsulfonic acid, Reychler's acid, d-10-Camphorsulfonic acid, d-Camphorsulfonic acid, β-Camphorsulfonic acid, (+)- |
| InChI Formula | InChI=1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m1/s1 |
| SMILES | CC1(C)[C@@H]2CC[C@@]1(CS(=O)(=O)O)C(=O)C2 |
| IUPAC Name | [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction