missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid, 97%

Product Code. p-7035043
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410096 250 mg 250mg
15400096 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410096 Supplier Thermo Scientific Alfa Aesar Supplier No. H52747.MD
Add to basket
This item is not returnable. View return policy

CAS: 96443-42-4 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96443-42-4
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD04972280
InChI Key FVUHGTQDOMGZOT-NTSWFWBYSA-N
Synonym 1s,3r-3-methoxycarbonyl cyclopentanecarboxylic acid, 1s,3r-3-methoxycarbonyl cyclopentane-1-carboxylic acid, 1r,3s-1,3-cyclopentanedicarboxylic acid 1-methyl ester, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxylic acid, cis-3-methoxycarbonyl cyclopentancarboxylic acid, 1s,3r-cis-3-carbomethoxy cyclopentane-1-carboxyl, 1,3-cyclopentanedicarboxylicacid,1-methyl ester, 1r,3s, 1r-cis-1,3-cyclopentanedicarboxylic acid monomethyl ester, 1,3-cyclopentanedicarboxylic acid, 1-methyl ester, 1r,3s
PubChem CID 10888375
IUPAC Name (1S,3R)-3-methoxycarbonylcyclopentane-1-carboxylic acid
SMILES COC(=O)C1CCC(C1)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material (1S,3R)-cis-3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.