missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-(-)-Camphoric acid, 98%

Product Code. 15480468
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15480468

Brand: Thermo Scientific Alfa Aesar H54042.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 560-09-8 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 560-09-8
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00064937
InChI Key LSPHULWDVZXLIL-QUBYGPBYSA-N
Synonym 1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid
PubChem CID 219463
IUPAC Name (1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
SMILES CC1(C(CCC1(C)C(=O)O)C(=O)O)C

Specifications

Melting Point 188°C to 190°C
Merck Index 141733
Formula Weight 200.23
Percent Purity 98%
Quantity 1 g
Optical Rotation -48° (c=10 in Ethanol)
Chemical Name or Material (1S,3R)-(-)-Camphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.