missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R,5R)-PIM447 dihydrochloride, MedChemExpress

Product Code. 16441799
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441799 1 mg 1mg
16480188 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441799 Supplier Medchem Express Supplier No. HY19322C

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress (1S,3R,5R)-PIM447 (dihydrochloride) an PIM inhibitor extracted from patent US 20100056576 A1, compound example 72, has IC50 values of 0.095 μM for Pim1, 0.522 μM for Pim2 and 0.369 μM for Pim3.

(1S,3R,5R)-PIM447 (dihydrochloride) an PIM inhibitor extracted from patent US 20100056576 A1, compound example 72, has IC50 values of 0.095 μM for Pim1, 0.522 μM for Pim2 and 0.369 μM for Pim3.

Specifications

Chemical Name or Material (1S,3R,5R)-PIM447 dihydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25Cl2F3N4O
Quantity 1 mg
Synonym (1S,3R,5R)-LGH447 dihydrochloride
Solubility Information DMSO : 33.33 mg/mL (64.92 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (24.35 mM; Need ultrasonic)
SMILES O=C(C1=NC(C2=C(F)C=CC=C2F)=C(F)C=C1)NC3=C([C@@H]4C[C@H](N)C[C@H](C)C4)C=CN=C3.Cl.Cl
Molecular Weight (g/mol) 513.38
Formula Weight 513.38
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.