missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(+)-3-Carene, TRC
Click to view available options
Quantity:
100 mg
Specifications
Specifications
| Chemical Name or Material | (1S)-(+)-3-Carene |
| CAS | 498-15-7 |
| Molecular Formula | C10 H16 |
| Quantity | 100 mg |
| Synonym | 3-Carene, (1S,6R)-(+)- (8CI), Bicyclo[4.1.0]hept-3-ene, 3,7,7-trimethyl-, (1S)-, (1S,6R)-3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene, (+)-3-Carene, (+)-Carene-3, (+)-δ3-Carene, (1S)-(+)-3-Carene, (1S)-3-Carene, (1S,6R)-3-Carene, (S)-(+)-3-Carene, Isodiprene |
| InChI Formula | InChI=1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9+/m1/s1 |
| SMILES | CC1=CC[C@@H]2[C@H](C1)C2(C)C |
| IUPAC Name | (1R,6S)-4,7,7-trimethylbicyclo[4.1.0]hept-3-ene |
| Molecular Weight (g/mol) | 136.23 |
| Formula Weight | 136.1252 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction