missing translation for 'onlineSavingsMsg'
Learn More

(1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic Acid 1-Methylethyl Ester (>90%), TRC

Product Code. 30137959
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137959 100 mg 100mg
30133056 50 mg 50mg
17900805 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137959 Supplier Toronto Research Chemicals Supplier No. TRCD474855100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1353274-25-5
Molecular Formula C19 H21 N O2
Molecular Weight (g/mol) 295.38
Synonym (1S)-3,4-Dihydro-1-phenyl-2(1H)-Isoquinolinecarboxylic acid 1-methylethyl ester, Solifenacin Succinate Imp. B (EP)
IUPAC Name propan-2-yl (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
SMILES CC(C)OC(=O)N1CCc2ccccc2[C@@H]1c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H21NO2/c1-14(2)22-19(21)20-13-12-15-8-6-7-11-17(15)18(20)16-9-4-3-5-10-16/h3-11,14,18H,12-13H2,1-2H3/t18-/m0/s1
Formula Weight 295.1572
Chemical Name or Material (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic acid 1-Methylethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.