missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 99%, Thermo Scientific™

Product Code. 11363906
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11363906 Supplier Thermo Scientific Alfa Aesar Supplier No. L17018.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(1S,2S)-(+)-2-Benzyloxycyclopentylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. It is sensitive to air and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181657-57-8
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD01075751
InChI Key JIMSXLUBRRQALI-RYUDHWBXSA-N
PubChem CID 11908836
IUPAC Name (1S,2S)-2-phenylmethoxycyclopentan-1-amine
SMILES C1CC(C(C1)OCC2=CC=CC=C2)N

Specifications

Melting Point 23°C
Density 0.983
Boiling Point 90°C to 93°C (0.23 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5305
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Synonym 1s,2s-1-amino-2-benzyloxycyclopentane, 1s,2s-2-benzyloxy cyclopentanamine, 1s,2s-2-benzyloxycyclopentylamine, 1s,2s-+-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxy cyclopentan-1-amine, 1s,25-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxycyclopentanamine, 1s, 2s-2-benzyloxycyclopentylamine, 1s,2s-2-benzyloxy-cyclopentylamine, 1s,2s-2-benzyloxycyclopentyl-amine
Solubility Information Not miscible or difficult to mix in water.
Optical Rotation +57.5° (Neat)
Formula Weight 191.27
Percent Purity ≥99%
Assay ee 99%
Chemical Name or Material (1S,2S)-(+)-2-Benzyloxycyclopentylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.